Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane, extra pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass Bottle
Plastic drum
110-82-7
C6H12
84.16
MFCD00003814
XDTMQSROBMDMFD-UHFFFAOYSA-N
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
8078
CHEBI:29005
cyclohexane
C1CCCCC1
110-82-7
C6H12
84.16
MFCD00003814
XDTMQSROBMDMFD-UHFFFAOYSA-N
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
8078
CHEBI:29005
cyclohexane
C1CCCCC1
Assay Percent Range 99.5%
Linear Formula C6H12
Formula Weight 84.15
Physical Form Liquid
Percent Purity 99.5%
Grade Extra Pure
Density 0.7700g/mL
Infrared Spectrum Authentic
Packaging Glass Bottle
Refractive Index 1.4250 to 1.4270
Residue after Evaporation 0.005% max.
Solubility Solubility in water: insoluble. Other solubilities: soluble in methanol (57 g/100 mL at 20°C), ethanol, , diethyl ether, acetone, benzene, ccl4.
Specific Gravity 0.77
Water 0.01% max.
Flash Point −18°C
Viscosity 0.94 mPa.s (20°C)
Boiling Point 81.0°C
Color Undesignated
Melting Point 6.5°C
Quantity 2.5L
Chemical Name or Material Cyclohexane
Show More Show Less

GHS H Statement
May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. Very toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Danger