Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane, For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass Bottle
110-82-7
C6H12
84.162
XDTMQSROBMDMFD-UHFFFAOYSA-N
hexamethylene,hexahydrobenzene,hexanaphthene,cyclohexan,cykloheksan,cicloesano,cyclohexaan,benzene, hexahydro,benzenehexahydride,polycyclohexane
8078
CHEBI:29005
cyclohexane
C1CCCCC1
110-82-7
C6H12
84.162
XDTMQSROBMDMFD-UHFFFAOYSA-N
hexamethylene,hexahydrobenzene,hexanaphthene,cyclohexan,cykloheksan,cicloesano,cyclohexaan,benzene, hexahydro,benzenehexahydride,polycyclohexane
8078
CHEBI:29005
cyclohexane
C1CCCCC1

For gas chromatography ECD and FID SupraSolv™

UN Number 1145
Solubility Information Soluble in water: 55mg/L at 20°C
Formula Weight 84.16
Physical Form Liquid
Description SupraSolv™ ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv™ solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.
Density 0.78g/cm3 at 20°C
Ignition Point 260°C (500°F)
Odor Charactristic
Packaging Glass Bottle
Refractive Index 1.42
Storage Information 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.
Vapor Pressure 103 hPa (20°C)
Flash Point −18°C (−0.4°F)
Boiling Point 81°C
Color Colorless
Melting Point 6°C
Quantity 1L
Chemical Name or Material Cyclohexane
Show More Show Less

H304: May be fatal if swallowed and enters airways.

H315: Causes skin irritation.

H225: Highly flammable liquid and vapour.