Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane, HPLC Grade, 99.5%, Spectrum™

Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
110-82-7
110-82-7
CAS Max % 1
Appearance Clear
Percent Purity 0.995
Grade High Purity Solvent
Packaging Amber Glass Bottle
Residue after Evaporation 3 ppm
Water 0.0002
Quantity 1 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.