Cyclohexane, HPLC Grade, 99% min.; Alfa Aesar™

Manufacturer: Alfa Aesar 22919K2

Quantity 1L

 View more versions of this product

Catalog No. AA22919K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 110-82-7
    Physical Form Liquid
    Assay Percent Range 99% min.
    Molecular Formula C6H12
    Formula Weight 84.16
    MDL Number MFCD00003814
    Chemical Name or Material Cyclohexane
    Grade HPLC Grade

    Specifications

    Formula Weight 84.16
    Quantity 1L
    Color Colorless
    DOT Information DOT Class: 3; Packing Group: II
    Health Hazard 2 Keep container in a well ventilated place. Keep away from sources of ignition - No Smoking. Avoid contact with eyes. Take precautionary measures against static discharges. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets If swallowed, do not induce vomiting: seek medical advice immediately & show this container or label.
    EINECS Number 203-806-2
    Physical Form Liquid
    Impurities Maximum level of impurities: Evaporation residue 3ppm, Chlorinated pesticides 5ppt, H2O 0.02%.
    Odor Pungent
    Vapor Pressure 104 hPa (78mm Hg) at 20°C
    Appearance Colorless liquid
    Assay Percent Range 99% min.
    Molecular Formula C6H12
    MDL Number MFCD00003814
    CAS 110-82-7
    Health Hazard 1 H225-H304-H400-H410-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Causes skin irritation. May cause drowsiness or dizziness.
    Chemical Name or Material Cyclohexane
    Grade HPLC Grade

    Keep container in a well ventilated place. Keep away from sources of ignition - No Smoking. Avoid contact with eyes. Take precautionary measures against static discharges. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets If swallowed, do not induce vomiting: seek medical advice immediately & show this container or label.

    H225-H304-H400-H410-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Causes skin irritation. May cause drowsiness or dizziness.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit