Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane, PestiSolv(R), 99%7, Spectrum™

Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
110-82-7
110-82-7
CAS Max % 1
Percent Purity 0.997
Grade High Purity Solvent
Packaging Amber Glass Bottle
Residue after Evaporation 5 ppm
Water 0.0002
Quantity 1 L

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.