Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane, Reagent, ACS, 99%, Spectrum™

Click to view available options
Quantity:
1 L
100 mL
1625 lbs.
20 L
200 L
4 L
4 x 4 L
500 mL
6 x 1 L
6 x 500 mL
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Steel Can
110-82-7
C6H12
84.16
XDTMQSROBMDMFD-UHFFFAOYSA-N
cyclohexane
C1CCCCC1
110-82-7
C6H12
84.16
XDTMQSROBMDMFD-UHFFFAOYSA-N
cyclohexane
C1CCCCC1

C1455, 110-82-7, C6H12

Spectrum™ Cyclohexane, Reagent, ACS is used as a nonpolar solvent, in the calibration of differential scanning calorimetry and other industrial applications including a chemical intermediate. The distinctive detergent-like odor of cyclohexane is reminiscent of cleaning product and is sometimes used in cleaning products as a fragrance enhancer.
CAS Max % 1
UN Number UN1145
Percent Purity 99%
Packaging Poly Drum
Residue after Evaporation 2e-005
Water 0.0002
Quantity 1625 lbs.