Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane, Spectrophotometric Grade, ACS, 99%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle
110-82-7
110-82-7
CAS Max % 1
Appearance Clear
Percent Purity 0.99
Grade Spectrophotometric
Packaging Amber Glass Bottle
Residue after Evaporation 2e-05
Water 0.0002
Quantity 4 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.