Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanecarboxylic Acid, 98+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
1kg
Packaging:
Glass bottle
98-89-5
C7H12O2
128.17
MFCD00001461
NZNMSOFKMUBTKW-UHFFFAOYSA-N
hexahydrobenzoic acid,carboxycyclohexane,cyclohexanoic acid,cyclohexylcarboxylic acid,cyclohexylmethanoic acid,cyclohexylformic acid,benzoic acid, hexahydro,cyclohexancarbonsaeure,cyclohexane-1-carboxylate,cyclohexanecarboxylicacid
7413
CHEBI:36096
cyclohexanecarboxylic acid
C1CCC(CC1)C(=O)O
98-89-5
C7H12O2
128.17
MFCD00001461
NZNMSOFKMUBTKW-UHFFFAOYSA-N
hexahydrobenzoic acid,carboxycyclohexane,cyclohexanoic acid,cyclohexylcarboxylic acid,cyclohexylmethanoic acid,cyclohexylformic acid,benzoic acid, hexahydro,cyclohexancarbonsaeure,cyclohexane-1-carboxylate,cyclohexanecarboxylicacid
7413
CHEBI:36096
cyclohexanecarboxylic acid
C1CCC(CC1)C(=O)O
Quantity 25g
Flash Point >112°C
Specific Gravity 1.03
Infrared Spectrum Authentic
Packaging Glass bottle
Water 0.4% max.
Density 1.0300g/mL
Boiling Point 232.0°C to 233.0°C
Refractive Index 1.4600 to 1.4620
Melting Point 30.0°C to 32.0°C
Color White
Assay Percent Range 98%
Linear Formula C6H11CO2H
Beilstein 09, 7
Merck Index 15, 2717
Solubility Information Solubility in water: 0.201g/100mL (15 c). Other solubilities: soluble in alcohol and ether
Formula Weight 128.17
Physical Form Crystalline Solid
Percent Purity ≥98%
Chemical Name or Material Cyclohexanecarboxylic acid, 98+%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 202-711-3

RTECSNumber : GU8370000

TSCA : TSCA