Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanemethylamine , ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3218-02-8
C7H15N
113.20
MFCD00001520
AVKNGPAMCBSNSO-UHFFFAOYSA-N
trans-2,3-dibromo-2-butene-1,4-diol,2,3-dibromo-2-butene-1,4-diol,e-2,3-dibromobut-2-ene-1,4-diol,2e-2,3-dibromobut-2-ene-1,4-diol,e-2,3-dibromo-2-butene-1,4-diol,2,3-dibromobutene-1,4-diol,ccris 4781,2-butene-1,4-diol, 2,3-dibromo-, 2e,2-butene-1,4-diol, 2,3-dibromo,trans-2,3-dibromo-2-buten-1,4-diol
641240
(E)-2,3-dibromobut-2-ene-1,4-diol
NCC1CCCCC1
3218-02-8
C7H15N
113.20
MFCD00001520
AVKNGPAMCBSNSO-UHFFFAOYSA-N
trans-2,3-dibromo-2-butene-1,4-diol,2,3-dibromo-2-butene-1,4-diol,e-2,3-dibromobut-2-ene-1,4-diol,2e-2,3-dibromobut-2-ene-1,4-diol,e-2,3-dibromo-2-butene-1,4-diol,2,3-dibromobutene-1,4-diol,ccris 4781,2-butene-1,4-diol, 2,3-dibromo-, 2e,2-butene-1,4-diol, 2,3-dibromo,trans-2,3-dibromo-2-buten-1,4-diol
641240
(E)-2,3-dibromobut-2-ene-1,4-diol
NCC1CCCCC1
Boiling Point 161°C to 163°C
Flash Point 43°C
Specific Gravity 0.87
Refractive Index 1.463
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
Density 0.8700g/mL
Color Colorless to Yellow
Assay Percent Range 97%
Linear Formula C6H11CH2NH2
Beilstein 12, 12
Formula Weight 113.2
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Cyclohexanemethylamine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger