Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanone, ≥99%, Alfa Aesar™

Click to view available options
Quantity:
100mL
500mL
2500mL
108-94-1
C6H10O
98.145
MFCD00001625
JHIVVAPYMSGYDF-UHFFFAOYSA-N
ketohexamethylene,pimelic ketone,sextone,cyclohexyl ketone,nadone,anone,anon,cyclohexanon,hytrol o,hexanon
7967
CHEBI:17854
cyclohexanone
C1CCC(=O)CC1
108-94-1
C6H10O
98.145
MFCD00001625
JHIVVAPYMSGYDF-UHFFFAOYSA-N
ketohexamethylene,pimelic ketone,sextone,cyclohexyl ketone,nadone,anone,anon,cyclohexanon,hytrol o,hexanon
7967
CHEBI:17854
cyclohexanone
C1CCC(=O)CC1
Applications
Cyclohexanone is used in the preparation of synthetic polymer such as nylon 6,6 and nylon 6. It acts as a solvent in organic synthesis. Its derivatives are used for the synthesis of dyes, rubber chemicals, plasticizers and pharmaceuticals. Further, it is involved in the preparation of cyclohexanone oxime, which rearranges to give caprolactam in the presence of sulfuric acid catalyst.

Solubility
Miscible with diethyl ether, acetone and alcohol. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, red metals, lead, amines, sulfuric acid, nitric acid, strong acids, aliphatic amines, rubber, resins and alkalies.
Assay Percent Range ≥99%
UN Number UN1915
Beilstein 385735
Merck Index 14,2726
Solubility Information Miscible with diethyl ether,acetone and alcohol. Slightly miscible with water.
Formula Weight 98.15
Physical Form Liquid
Percent Purity ≥99%
Density 0.948
Refractive Index 1.4507
Flash Point 47°C (116°F)
Boiling Point 154°C to 156°C
Melting Point -26°C
Quantity 100mL
Chemical Name or Material Cyclohexanone
Show More Show Less

GHS H Statement
H226-H332
Flammable liquid and vapour.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c

H226-H302+H312+H332-H315-H318-H335-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYCLOHEXANONE

EINECSNumber : 203-631-1

RTECSNumber : GW1050000

TSCA : Yes

Recommended Storage : Ambient temperatures