Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanone, ACS, 99%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
108-94-1
108-94-1
CAS Max % 1
Percent Purity 0.99
Grade ACS
Packaging Amber Glass Bottle
Residue after Evaporation 0.0005
Water 0.0005
Quantity 1 L

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.