Alfa Aesar™ Cyclohexanone cyanohydrin, 98%

Manufacturer: Alfa Aesar™ H2669903

Quantity 1g

 View more versions of this product

Catalog No. AAH2669903

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 931-97-5
    Molecular Formula C7H11NO
    Formula Weight 125.17
    MDL Number MFCD00003818
    UN Number UN3439
    Synonym 1-Hydroxycyclohexanecarbonitrile
    Chemical Name or Material Cyclohexanone cyanohydrin

    Specifications

    Formula Weight 125.17
    Quantity 1g
    TSCA TSCA
    Molecular Formula C7H11NO
    MDL Number MFCD00003818
    CAS 931-97-5
    UN Number UN3439
    Synonym 1-Hydroxycyclohexanecarbonitrile
    Health Hazard 1 Danger
    Boiling Point 132°C (19mmHg)
    Melting Point 34° to 36°C
    Flash Point 118°C (244°F)
    Recommended Storage 2° to 8°C
    RTECS Number GU7710000
    Health Hazard 2 GHS H Statement
    H300
    Fatal if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material Cyclohexanone cyanohydrin
    Assay 98%

    GHS H Statement
    H300
    Fatal if swallowed.

    2° to 8°C

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit