Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cyclohexanone oxime, 97%

Click to view available options
Quantity:
100g
500g
100-64-1
C6H11NO
113.16
MFCD00001660
VEZUQRBDRNJBJY-UHFFFAOYSA-N
cyclohexanone oxime,cyclohexanone, oxime,antioxidant d,hydroxyimino cyclohexane,unii-2u60l00cgf,ccris 1383,dsstox_cid_1842,dsstox_rid_76361,dsstox_gsid_21842,cyclohexanoneoxime
7517
N-cyclohexylidenehydroxylamine
C1CCC(=NO)CC1
100-64-1
C6H11NO
113.16
MFCD00001660
VEZUQRBDRNJBJY-UHFFFAOYSA-N
cyclohexanone oxime,cyclohexanone, oxime,antioxidant d,hydroxyimino cyclohexane,unii-2u60l00cgf,ccris 1383,dsstox_cid_1842,dsstox_rid_76361,dsstox_gsid_21842,cyclohexanoneoxime
7517
N-cyclohexylidenehydroxylamine
C1CCC(=NO)CC1
Flash Point 103°C (217°F)
Melting Point 87°C to 91°C
Boiling Point 206°C to 210°C
Quantity 500g
Beilstein 1616769
Formula Weight 113.16
Percent Purity 97%
Chemical Name or Material Cyclohexanone oxime

GHS H Statement
H301
Toxic if swallowed.

P210-P240-P241-P260-P264b-P270-P280-P301+P312-P305+P351+P338-P314-P330-P370+P378q-P501c

H228-H302-H319-H373

DOTInformation : Hazard Class: 4.1; Packaging Group: III

EINECSNumber : 202-874-0

RTECSNumber : GW1925000

TSCA : Yes

Recommended Storage : Ambient temperatures