Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanone, Technical, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
55 gal
Packaging:
Amber Glass Bottle
Drum
Steel Can
108-94-1
108-94-1
CAS Max % 1
Appearance Clear, Colorless
Grade Technical
Packaging Steel Can
Quantity 20 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.