Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexene, 98%, Spectrum™

Click to view available options
Quantity:
25 mL
250 mL
1 L
200 L
Packaging:
Amber Glass Bottle
Drum
110-83-8
110-83-8
CAS Max % 1
Percent Purity 0.98
Grade Reagent
Packaging Amber Glass Bottle
Quantity 250 mL

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.