Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Cycloheximide, >98%, Calbiochem™
Click to view available options
Quantity:
1 g
Packaging:
Alu drum
Description
- Antifungal antibiotic that inhibits protein synthesis in eukaryotes but not in prokaryotes
- Interacts directly with translocase enzyme, interfering with the translocation step
- Inhibits cell-free protein synthesis in eukaryotes
- Competitively inhibits hFKBP12 (Ki = 3.4 μM)
- Triggers apoptosis in HL-60 cells, T cell hybridomas, Burkitt’s lymphoma cells, and a variety of other cell types including rodent macrophages
- Inhibits DNA cleavage in rat thymocytes treated with Thapsigargin, methylprednisolone, and Ionomycin
- Rapidly destroyed in alkaline solutions.
- Also available as a 300 mM solution in DMSO
Recommended Storage
+15°C to +30°C, Following reconstitution, aliquot and freeze (-20°C) for long term storage or refrigerate (4°C) for short term storage. Stock solutions are stable for up to 6 weeks at 4°C, pH 3-5.
Specifications
Packaging | Alu drum |
Quantity | 1 g |
Merck Index | 14, 2728 |
Solubility Information | Soluble in Chloroform, Ethanol, and Methanol |
Percent Purity | ≥98% |
Physical Form | Solid |
Chemical Name or Material | Cycloheximide, High Purity |
Grade | By HPLC |
Safety and Handling
Hazard Category | Highly Toxic & Carcinogenic / Teratogenic |
Hazard Statement | R: 28-40-51/53-61 Very toxic if swallowed. Limited evidence of a carcinogenic effect. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. May cause harm to the unborn child. |
Precautionary Statement | S: 45-53-61 In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid exposure - obtain special instructions before use. Avoid release to the environment. |
RTECSNumber | MA4375000 |
storageNote1 | Following reconstitution, aliquot and freeze (-20°C) for long term storage or refrigerate (4°C) for short term storage. Stock solutions are stable for up to 6 weeks at 4°C, pH 3-5. |
Recommended Storage | +15°C to +30°C; Ok to freeze; |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction