Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cyclohexyl hexanoate, 99%

Click to view available options
Quantity:
25g
100g
6246-05-5
C4H9NO
87.12
MFCD00046356
KTHZBRAXOLUNBN-UHFFFAOYSA-N
3-oxetanemethanamine,3-aminomethyl-oxetane,3-aminomethyloxetane,1-oxetan-3-yl methanamine,c-oxetan-3-yl-methylamine,oxetan-3-ylmethamine,oxetane-3-methanamine,n-methyl-3-oxetanamne,oxetane-3-ylmethanamine,pubchem23208
46840007
oxetan-3-ylmethanamine
C1C(CO1)CN
6246-05-5
C4H9NO
87.12
MFCD00046356
KTHZBRAXOLUNBN-UHFFFAOYSA-N
3-oxetanemethanamine,3-aminomethyl-oxetane,3-aminomethyloxetane,1-oxetan-3-yl methanamine,c-oxetan-3-yl-methylamine,oxetan-3-ylmethamine,oxetane-3-methanamine,n-methyl-3-oxetanamne,oxetane-3-ylmethanamine,pubchem23208
46840007
oxetan-3-ylmethanamine
C1C(CO1)CN
Quantity 100g
Formula Weight 198.31
Percent Purity 99%
Chemical Name or Material Cyclohexyl hexanoate