Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexyl iodide, 98%, stabilized, Thermo Scientific Chemicals

Catalog Number ac218490250
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
626-62-0
C6H11I
210.06
MFCD00003826
FUCOMWZKWIEKRK-UHFFFAOYSA-N
cyclohexane, iodo, cyclohexyl iodide, cyclohexyliodide, iodo-cyclohexane, 1-iodocyclohexane, heptynyltrimethylsilane, iodocyclohexane, acmc-1att4, c-c6h11i
12289
iodocyclohexane
IC1CCCCC1
626-62-0
C6H11I
210.06
MFCD00003826
FUCOMWZKWIEKRK-UHFFFAOYSA-N
cyclohexane, iodo, cyclohexyl iodide, cyclohexyliodide, iodo-cyclohexane, 1-iodocyclohexane, heptynyltrimethylsilane, iodocyclohexane, acmc-1att4, c-c6h11i
12289
iodocyclohexane
IC1CCCCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Density 1.6200g/mL
Boiling Point 80.0°C to 81.0°C (20.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5470 to 1.5490
Linear Formula C6H11I
Quantity 25 mL
Beilstein 05, 25
Specific Gravity 1.62
Formula Weight 210.06
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclohexyl iodide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-957-8

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit