Alfa Aesar™ Cyclohexyl isocyanate, 98%

Manufacturer: Alfa Aesar™ B2305914

Quantity 25g

 View more versions of this product

Catalog No. AAB2305914

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 3173-53-3
    Molecular Formula C7H11NO
    Formula Weight 125.17
    MDL Number MFCD00003840
    UN Number UN2488
    Beilstein 507983
    Chemical Name or Material Cyclohexyl isocyanate

    Specifications

    Formula Weight 125.17
    TSCA TSCA
    Odor Pungent
    MDL Number MFCD00003840
    UN Number UN2488
    Beilstein 507983
    Health Hazard 2 GHS H Statement
    H301-H311-H330-H334-H318-H226-H315-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Fatal if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes serious eye damage.
    Flammable liquid and vapour.
    Causes skin irritation.
    May cause respiratory irritation.
    Flash Point 48°C (118°F)
    Boiling Point 168° to 170°C
    Melting Point ≈−80°C
    Quantity 25g
    Molecular Formula C7H11NO
    CAS 3173-53-3
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: CYCLOHEXYL ISOCYANATE
    RTECS Number NQ8650000
    Chemical Name or Material Cyclohexyl isocyanate
    Assay 98%

    GHS H Statement
    H301-H311-H330-H334-H318-H226-H315-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Fatal if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes serious eye damage.
    Flammable liquid and vapour.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit