Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cyclohexyl isothiocyanate, 98%

Manufacturer:  Alfa Aesar™ A1558514

Quantity 25g

 View more versions of this product

Catalog No. AAA1558514

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1122-82-3
MFCD00003841
774525
141.24
UN2922
Cyclohexyl isothiocyanate

Specifications

1.033g/mL
219°C
1122-82-3
1.5380
141.24
UN2922
Cyclohexyl isothiocyanate
Moisture Sensitive
25g
Pungent
Liquid
MFCD00003841
774525
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P305+P351+P338-P308+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or concerned: Get medical advice/attention.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

RTECSNumber : NX8480000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit