Alfa Aesar™ Cyclohexyl isothiocyanate, 98%

Manufacturer: Alfa Aesar™ A1558506

Quantity 5g

 View more versions of this product

Catalog No. AAA1558506

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1122-82-3
    Formula Weight 141.24
    MDL Number MFCD00003841
    UN Number UN2922
    Beilstein 774525
    Chemical Name or Material Cyclohexyl isothiocyanate

    Specifications

    Formula Weight 141.24
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P308+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or concerned: Get medical advice/attention.
    MDL Number MFCD00003841
    Beilstein 774525
    Health Hazard 2 GHS H Statement
    H331-H314-H318-H302-H312
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Boiling Point 219°C
    Refractive Index 1.5380
    Physical Form Liquid
    UN Number UN2922
    Health Hazard 1 Danger
    Density 1.033g/mL
    Sensitivity Moisture Sensitive
    TSCA TSCA
    Odor Pungent
    CAS 1122-82-3
    DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
    RTECS Number NX8480000
    Quantity 5g
    Chemical Name or Material Cyclohexyl isothiocyanate
    Assay 98%

    GHS H Statement
    H331-H314-H318-H302-H312
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P280-P305+P351+P338-P308+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or concerned: Get medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit