Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cyclohexyl phenyl ketone, 98%

Click to view available options
Quantity:
5g
25g
100g
712-50-5
C13H16O
188.27
MFCD00001467
BMFYCFSWWDXEPB-UHFFFAOYSA-N
cyclohexyl phenyl ketone,cyclohexyl phenyl methanone,benzoylcyclohexane,cyclohexylphenylketone,benzoyl cyclohexane,phenyl cyclohexyl ketone,methanone, cyclohexylphenyl,cyclohexyl-phenyl-methanone,usaf kf-3,cyclohexylphenyl ketone
12837
cyclohexyl(phenyl)methanone
C1CCC(CC1)C(=O)C2=CC=CC=C2
712-50-5
C13H16O
188.27
MFCD00001467
BMFYCFSWWDXEPB-UHFFFAOYSA-N
cyclohexyl phenyl ketone,cyclohexyl phenyl methanone,benzoylcyclohexane,cyclohexylphenylketone,benzoyl cyclohexane,phenyl cyclohexyl ketone,methanone, cyclohexylphenyl,cyclohexyl-phenyl-methanone,usaf kf-3,cyclohexylphenyl ketone
12837
cyclohexyl(phenyl)methanone
C1CCC(CC1)C(=O)C2=CC=CC=C2
Melting Point 52°C to 57°C
Quantity 100g
Flash Point >110°C (230°F)
Boiling Point 165°C to 168°C (20mmHg)
Beilstein 2046712
Formula Weight 188.27
Percent Purity 98%
Chemical Name or Material Cyclohexyl phenyl ketone

RTECSNumber : OB1760000

TSCA : TSCA