Alfa Aesar™ Cyclohexylacetylene, 98%

Manufacturer: Alfa Aesar™ H53418-06

Quantity 5g

 View more versions of this product

Catalog No. AAH5341806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 931-48-6
    Molecular Formula C8H12
    Formula Weight 108.18g/mol
    MDL Number MFCD00001513
    UN Number UN3295
    Beilstein 1815535
    Synonym Ethynylcyclohexane
    Chemical Name or Material Cyclohexylacetylene

    Specifications

    Quantity 5g,5g
    Boiling Point 130° to 132°C
    Flash Point 18°C (64°F)
    CAS 931-48-6
    Molecular Formula C8H12
    Formula Weight 108.18g/mol
    MDL Number MFCD00001513
    UN Number UN3295
    Beilstein 1815535
    Synonym Ethynylcyclohexane
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225
    Highly flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
    Chemical Name or Material Cyclohexylacetylene
    Assay 98%

    Description

    Applications
    Cyclohexylacetylene is used in the preparation of hydrido-vinylidene complexes of osmium, which finds vital intermediates in several homogeneous and heterogeneous catalytic reactions such as alkene olimerization, polymerization and Fischer-Tropsch synthesis.

    Solubility
    Immiscible with water.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H225
    Highly flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit