Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexylamine Hydrochloride 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
4998-76-9
C6H13N·HCl
135.63
MFCD00050540
ZJUGSKJHHWASAF-UHFFFAOYSA-N
cyclohexylamine hydrochloride,cyclohexanamine hydrochloride,cyclohexanamine, hydrochloride,aminocyclohexane hydrochloride,cyclohexylamine.hcl,unii-tn6vyf8thc,dsstox_cid_361,cyclohexanamine, hydrochloride 1:1,dsstox_rid_75539,dsstox_gsid_20361
2723770
cyclohexanamine;hydrochloride
C1CCC(CC1)N.Cl
4998-76-9
C6H13N·HCl
135.63
MFCD00050540
ZJUGSKJHHWASAF-UHFFFAOYSA-N
cyclohexylamine hydrochloride,cyclohexanamine hydrochloride,cyclohexanamine, hydrochloride,aminocyclohexane hydrochloride,cyclohexylamine.hcl,unii-tn6vyf8thc,dsstox_cid_361,cyclohexanamine, hydrochloride 1:1,dsstox_rid_75539,dsstox_gsid_20361
2723770
cyclohexanamine;hydrochloride
C1CCC(CC1)N.Cl
Packaging Plastic bottle
Melting Point 207.0°C
Infrared Spectrum Authentic
Color White to Beige
Boiling Point 306.0°C
Quantity 100g
Water 0.5% max. (K.F.)
Assay Percent Range 99%
Solubility Information Solubility in water: 83 g/100mL water (17°C). Other solubilities: soluble in most common organic solvents, practically insoluble in ethers
Formula Weight 135.63
Physical Form Crystalline Powder or Crystals
Percent Purity 99%
Chemical Name or Material Cyclohexylamine hydrochloride, 99%
Show More Show Less

GHS P Statement Causes serious eye irritation. Causes skin irritation. Harmful if swallowed. May cause respiratory irritation. Suspected of damaging fertility.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

HYGROSCOPIC

EINECSNumber : 225-661-4

RTECSNumber : GX1440000

TSCA : TSCA