Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexylboronic acid pinacol ester, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
87100-15-0
C12H23BO2
210.13
MFCD04038749
OUEVCDGYTKLNMJ-UHFFFAOYSA-N
cyclohexylboronic acid pinacol ester,1,3,2-dioxaborolane, 2-cyclohexyl-4,4,5,5-tetramethyl,cyclohexylboronic acid, pinacol ester,4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohexane,pubchem7924,cyclohexylboronic acid, pinacol eser,1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohexane
3668596
2-cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2CCCCC2
87100-15-0
C12H23BO2
210.13
MFCD04038749
OUEVCDGYTKLNMJ-UHFFFAOYSA-N
cyclohexylboronic acid pinacol ester,1,3,2-dioxaborolane, 2-cyclohexyl-4,4,5,5-tetramethyl,cyclohexylboronic acid, pinacol ester,4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohexane,pubchem7924,cyclohexylboronic acid, pinacol eser,1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohexane
3668596
2-cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2CCCCC2
Infrared Spectrum Authentic
Color Colorless to Yellow
Packaging Glass bottle
Quantity 1g
Assay Percent Range 96% min. (GC)
Formula Weight 210.13
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Cyclohexylboronic acid pinacol ester

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning