Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Cyclohexyldiphenylphosphine, 98%
Click to view available options
Quantity:
1g
5g
25g
100g
Specifications
Flash Point | >110°C (230°F) |
Melting Point | 58°C to 62°C |
Quantity | 1g |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 268.34 |
Physical Form | Solid |
Percent Purity | 98% |
Chemical Name or Material | Cyclohexyldiphenylphosphine |
Safety and Handling
P260-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P314-P330-P333+P313-P362-P501c
H302-H315-H317-H319-H373
EINECSNumber : 228-904-2
TSCA : No
Recommended Storage : Ambient temperatures