Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexylmagnesium chloride, 1.3M solution in THF/toluene, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
931-51-1,109-99-9,108-88-3
C6H11ClMg
142.91
MFCD00003816
DEDWWARPYWCXMG-UHFFFAOYSA-M
cyclohexylmagnesium chloride,magnesium, chlorocyclohexyl,magnesium,chlorocyclohexyl,cyclohexyl magnesium chloride,chloro cyclohexyl magnesium,chlorocyclohexylmagnesium,cyclohexylmagnesiumchloride,unii-oag686rdsu,cyclohexylmagnesium chloride solution, 2.0 m in diethyl ether,cyclohexylmagnesium cloride
13605
magnesium;cyclohexane;chloride
C1CC[CH-]CC1.[Mg+2].[Cl-]
931-51-1,109-99-9,108-88-3
C6H11ClMg
142.91
MFCD00003816
DEDWWARPYWCXMG-UHFFFAOYSA-M
cyclohexylmagnesium chloride,magnesium, chlorocyclohexyl,magnesium,chlorocyclohexyl,cyclohexyl magnesium chloride,chloro cyclohexyl magnesium,chlorocyclohexylmagnesium,cyclohexylmagnesiumchloride,unii-oag686rdsu,cyclohexylmagnesium chloride solution, 2.0 m in diethyl ether,cyclohexylmagnesium cloride
13605
magnesium;cyclohexane;chloride
C1CC[CH-]CC1.[Mg+2].[Cl-]
Specific Gravity 0.95
Flash Point −21°C
Density 0.9500g/mL
Packaging AcroSeal™ Glass bottle
Quantity 100mL
Color Brown
Linear Formula (C6H11)MgCl
Solubility Information Solubility in water: reacts violently with water
Formula Weight 142.91
Physical Form Solution
Chemical Name or Material Cyclohexylmagnesium chloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Suspected of damag

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 213-237-1