Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexylmethylmagnesium bromide, 0.25M solution in THF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
Packaging:
AcroSeal™ Glass Bottle
35166-78-0,109-99-9
C7H13BrMg
201.39
UFQSTBGEWZUCBQ-UHFFFAOYSA-M
cyclohexylmethylmagnesium bromide,cyclohexylmethyl magnesium bromide solution,zbciogfgzrnnec-uhfffaoysa-m,magnesium,bromo cyclohexylmethyl,bromo cyclohexylmethyl magnesium,cyclohexylmethylmagnesium bromide 0.25 m in tetrahydrofuran,cyclohexylmethylmagnesium bromide, 0.25m in tetrahydrofuran
10932511
magnesium;methanidylcyclohexane;bromide
[CH2-]C1CCCCC1.[Mg+2].[Br-]
35166-78-0,109-99-9
C7H13BrMg
201.39
UFQSTBGEWZUCBQ-UHFFFAOYSA-M
cyclohexylmethylmagnesium bromide,cyclohexylmethyl magnesium bromide solution,zbciogfgzrnnec-uhfffaoysa-m,magnesium,bromo cyclohexylmethyl,bromo cyclohexylmethyl magnesium,cyclohexylmethylmagnesium bromide 0.25 m in tetrahydrofuran,cyclohexylmethylmagnesium bromide, 0.25m in tetrahydrofuran
10932511
magnesium;methanidylcyclohexane;bromide
[CH2-]C1CCCCC1.[Mg+2].[Br-]
Specific Gravity 0.966
Color Brown to Colorless
Density 0.9660g/mL
Flash Point −20°C
Quantity 100mL
Packaging AcroSeal™ Glass Bottle
Solubility Information Solubility in water: reacts
Formula Weight 201.39
Physical Form Solution
Chemical Name or Material Cyclohexylmethylmagnesium bromide

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapour.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger