Alfa Aesar™ Cyclohexylurea, 98%

Manufacturer: Alfa Aesar™ B2491536

Quantity 500g

 View more versions of this product

Catalog No. AAB2491536

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 698-90-8
    Molecular Formula C7H14N2O
    Formula Weight 142.2
    MDL Number MFCD00014286
    Beilstein 636914
    Chemical Name or Material Cyclohexylurea

    Specifications

    Formula Weight 142.2
    Molecular Formula C7H14N2O
    MDL Number MFCD00014286
    CAS 698-90-8
    Beilstein 636914
    RTECS Number YS7564300
    Melting Point 191° to 195°C
    Quantity 500g
    Chemical Name or Material Cyclohexylurea
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit