Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Cyclooctene, 95%, stabilized, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
931-87-3
C8H14
110.2
URYYVOIYTNXXBN-UPHRSURJSA-N
cis-cyclooctene,z-cyclooctene,cyclooctene, z,cyclooctene, 1z,cyclooct-1-ene,e-cyclooctene,1-cyclooctene #,1z-cyclooctene,7z-cyclooctene,ar,e-cyclooctene
638079
cyclooctene
C1CCCC=CCC1
931-87-3
C8H14
110.2
URYYVOIYTNXXBN-UPHRSURJSA-N
cis-cyclooctene,z-cyclooctene,cyclooctene, z,cyclooctene, 1z,cyclooct-1-ene,e-cyclooctene,1-cyclooctene #,1z-cyclooctene,7z-cyclooctene,ar,e-cyclooctene
638079
cyclooctene
C1CCCC=CCC1
Refractive Index 1.4680 to 1.4710
Packaging Glass bottle
Stabilizer 100 to 200ppm irganox 1076 FD
Infrared Spectrum Authentic
Quantity 1L
Flash Point 25°C
Specific Gravity 0.84
Viscosity 1.89 mPa.s (20°C)
Boiling Point 145.0°C to 146.0°C
Melting Point -16.0°C
Color Colorless to Brown
Density 0.8400g/mL
Assay Percent Range 94% min. (GC)
Linear Formula C8H14
Solubility Information Solubility in water: <0.1g/L (20°C)
Formula Weight 110.2
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material cis-Cyclooctene
Show More Show Less

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.

GHS P Statement
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
Store in a well

GHS Signal Word: Danger

EINECSNumber : 213-243-4