Alfa Aesar™ Cyclopentadienyltitanium trichloride

Manufacturer: Alfa Aesar™ 7713403

Quantity 1g

 View more versions of this product

Catalog No. AA7713403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1270-98-0
    Molecular Formula C5H5Cl3Ti
    Formula Weight 219.35
    MDL Number MFCD00003724
    UN Number UN3261
    Synonym Trichlorocyclopentadienyltitanium
    Chemical Name or Material Cyclopentadienyltitanium trichloride

    Specifications

    Formula Weight 219.35
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00003724
    Color Yellow-orange
    Quantity 1g
    Sensitivity Moisture Sensitive
    CAS 1270-98-0
    Synonym Trichlorocyclopentadienyltitanium
    Melting Point 210°C (dec.)
    Health Hazard 1 Danger
    RTECS Number XR1927300
    Physical Form Crystalline
    Molecular Formula C5H5Cl3Ti
    UN Number UN3261
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material Cyclopentadienyltitanium trichloride

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit