Alfa Aesar™ Cyclopentadienylvanadium tetracarbonyl, 97+%

Manufacturer: Alfa Aesar™ 8112304

Quantity 2g

 View more versions of this product

Catalog No. AA8112304

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12108-04-2
    Molecular Formula C9H5O4V
    Formula Weight 228.08
    MDL Number MFCD00014259
    UN Number UN3466
    Synonym Tetracarbonylcyclopentadienylvanadium
    Chemical Name or Material Cyclopentadienylvanadium tetracarbonyl

    Specifications

    Formula Weight 228.08
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H315-H319-H335
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Sensitivity Air Sensitive
    Quantity 2g
    MDL Number MFCD00014259
    Synonym Tetracarbonylcyclopentadienylvanadium
    Melting Point 138°C
    Physical Form Crystalline
    Molecular Formula C9H5O4V
    CAS 12108-04-2
    UN Number UN3466
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
    Boiling Point 80° to 100°C (0.1mmHg) (subl.)
    Chemical Name or Material Cyclopentadienylvanadium tetracarbonyl
    Assay ≥97%

    GHS H Statement
    H300-H310-H330-H315-H319-H335
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit