Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentadienylzirconium Trichloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
34767-44-7
C5H5Cl3Zr
MFCD00015756
34767-44-7
C5H5Cl3Zr
MFCD00015756
Quantity 1g
Color White to Brown
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
Formula Weight 262.67
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material Cyclopentadienylzirconium trichloride

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger