Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentane, Honeywell Burdick & Jackson™

Click to view available options
Quantity:
200 L
Packaging:
Returnable Steel Drum
9253
CHEBI:23492
9253
CHEBI:23492

B&J Brand™, for HPLC, GC, pesticide residue analysis, and spectrophotometry, >:99.9%

Flash Point -7 °C
Packaging Returnable Steel Drum
Refractive Index 1.3930 - 1.3970
Quantity 200 L
Grade Multipurpose Solvents
Physical Form Liquid

Recommended Storage : Room Temperature