Cyclopentane, HPLC Grade, Alfa Aesar™

For HPLC and Spectrophotometry

Manufacturer: Alfa Aesar 44814K7

Quantity 4L

 View more versions of this product

Catalog No. AA44814K7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 287-92-3
    Physical Form Liquid
    Molecular Formula C5H10
    Formula Weight 70.14
    MDL Number MFCD00001356
    Chemical Name or Material Cyclopentane, Contains 75+% cyclopentane. Remainder consists of C5 and C6 hydrocarbons .
    Grade HPLC Grade

    Specifications

    Physical Form Liquid
    Formula Weight 70.14
    Color Clear colorless
    Quantity 4L
    Additional Information For HPLC and Spectrophotometry
    UN Number 1146
    Density 0.748g/cm3 at 20°C
    Flash Point −37°C (−34.6°F)
    Boiling Point 47° to 49°C
    Melting Point −95°C (−139°F)
    DOT Information DOT Class: 3, Packaging Group: II
    EINECS Number 206-016-6
    Ignition Point 380°C (716°F)
    Odor Petrol-like
    Vapor Pressure 398mm Hg at 20°C
    Molecular Formula C5H10
    MDL Number MFCD00001356
    CAS 287-92-3
    Health Hazard 1 H225-H412: Highly flammable liquid and vapour. Harmful to aquatic life with long lasting effects.
    Chemical Name or Material Cyclopentane, Contains 75+% cyclopentane. Remainder consists of C5 and C6 hydrocarbons .
    Grade HPLC Grade

    H225-H412: Highly flammable liquid and vapour. Harmful to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit