Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentanemethanol 96%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
Packaging:
Glass bottle
3637-61-4
C6H12O
100.16
MFCD00001384
ISQVBYGGNVVVHB-UHFFFAOYSA-N
cyclopentanemethanol,cyclopentyl-methanol,cyclopentylcarbinol,cyclopentylmethyl alcohol,cyclopentanmethanol,cylcopentanemethanol,cyclopentan-methanol,cyclopentyl methanol,cyclopentyl carbinol,cyclo-pentanemethanol
77195
cyclopentylmethanol
C1CCC(C1)CO
3637-61-4
C6H12O
100.16
MFCD00001384
ISQVBYGGNVVVHB-UHFFFAOYSA-N
cyclopentanemethanol,cyclopentyl-methanol,cyclopentylcarbinol,cyclopentylmethyl alcohol,cyclopentanmethanol,cylcopentanemethanol,cyclopentan-methanol,cyclopentyl methanol,cyclopentyl carbinol,cyclo-pentanemethanol
77195
cyclopentylmethanol
C1CCC(C1)CO
Boiling Point 162°C to 163°C
Flash Point 62°C
Density 0.9200g/mL
Specific Gravity 0.92
Refractive Index 1.4565 to 1.4585
Packaging Glass bottle
Quantity 5g
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula C5H9CH2OH
Beilstein 06, 9
Formula Weight 100.16
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Cyclopentanemethanol, 96%
Show More Show Less

EINECSNumber : 222-861-3