Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentanethiol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
1679-07-8
C5H10S
102.2
MFCD00001369
WVDYBOADDMMFIY-UHFFFAOYSA-N
cyclopentyl mercaptan,mercaptocyclopentane,cyclopentylthiol,fema no. 3262,cyclopentyl thiol,cyclopen-tanethiol,cyclopentane thiol,cyclopentylmercaptan,acmc-209dxb,cyclopentanethiol
15510
cyclopentanethiol
C1CCC(C1)S
1679-07-8
C5H10S
102.2
MFCD00001369
WVDYBOADDMMFIY-UHFFFAOYSA-N
cyclopentyl mercaptan,mercaptocyclopentane,cyclopentylthiol,fema no. 3262,cyclopentyl thiol,cyclopen-tanethiol,cyclopentane thiol,cyclopentylmercaptan,acmc-209dxb,cyclopentanethiol
15510
cyclopentanethiol
C1CCC(C1)S
Flash Point 25°C
Boiling Point 129°C to 131°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 5g
Assay Percent Range 97.5% min. (GC)
Formula Weight 102.2
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Cyclopentanethiol
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

EINECSNumber : 216-841-3