Alfa Aesar™ Cyclopentanone oxime, 97%

Manufacturer: Alfa Aesar™ B2496122

Quantity 100g

 View more versions of this product

Catalog No. AAB2496122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1192-28-5
    Molecular Formula C5H9NO
    Formula Weight 99.13
    MDL Number MFCD00001420
    Chemical Name or Material Cyclopentanone oxime

    Specifications

    Formula Weight 99.13
    TSCA TSCA
    MDL Number MFCD00001420
    Flash Point 92°C (198°F)
    Boiling Point 196°C
    Melting Point 54° to 58°C
    Quantity 100g
    Molecular Formula C5H9NO
    CAS 1192-28-5
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number GY5150000
    Chemical Name or Material Cyclopentanone oxime
    Assay 97%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit