Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Cyclopentanone oxime, 97%
Click to view available options
Quantity:
25g
100g
Specifications
Boiling Point | 196°C |
Flash Point | 92°C (198°F) |
Quantity | 25g |
Melting Point | 54°C to 58°C |
Formula Weight | 99.13 |
Percent Purity | 97% |
Chemical Name or Material | Cyclopentanone oxime |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 214-749-8
RTECSNumber : GY5150000
TSCA : Yes
Recommended Storage : Ambient temperatures