Alfa Aesar™ Cyclopentene, 97%

Manufacturer: Alfa Aesar™ A11027AP

Quantity 500mL

 View more versions of this product

Catalog No. AAA11027AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 142-29-0
    Molecular Formula C5H8
    Formula Weight 68.12
    MDL Number MFCD00001394
    UN Number UN2246
    Beilstein 635707
    Chemical Name or Material Cyclopentene

    Specifications

    Formula Weight 68.12
    Health Hazard 3 GHS P Statement
    P210-P280h-P273-P301+P330+P331-P315-P403
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
    Get immediate medical advice/attention.
    Store in a well-ventilated place
    TSCA TSCA
    Molecular Formula C5H8
    MDL Number MFCD00001394
    Recommended Storage 2°C to 8°C
    RTECS Number GY5950000
    UN Number UN2246
    Health Hazard 2 GHS H Statement
    H225-H304-H302-H312-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Harmful if swallowed.
    Harmful in contact with skin.
    May cause drowsiness or dizziness.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CYCLOPENTENE
    Flash Point −29°C (−20°F)
    CAS 142-29-0
    Health Hazard 1 Danger
    Boiling Point 44° to 45°C
    Melting Point −135°C
    Quantity 500mL
    Beilstein 635707
    Chemical Name or Material Cyclopentene
    Assay 97%

    Description

    Applications
    Cyclopentene is used in the production of cyclopentene oxide, ketamine, cyclopentyl phenol (disinfectant), aldehyde and cyclopentene glycol. It acts as a monomer and is utilized for the synthesis of plastics and in various chemical syntheses. It is also used in the investigation of [2+2] cycloaddition of diamond (001) surfaces with alkene.

    Solubility
    Miscible with ethanol, ether, benzene and organic solvents. Immiscible with water.

    Notes
    Store in cool place. Air sensitive. Keep container tightly closed in a dry and well-ventilated place.

    GHS H Statement
    H225-H304-H302-H312-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Harmful if swallowed.
    Harmful in contact with skin.
    May cause drowsiness or dizziness.

    2°C to 8°C

    GHS P Statement
    P210-P280h-P273-P301+P330+P331-P315-P403
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
    Get immediate medical advice/attention.
    Store in a well-ventilated place

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit