Learn More
Sigma Organic Chemistry CyclopentY isothiocyana | 100mg | 33522-03-1 | MFCD00040877
Supplier: Sigma Organic Chemistry CDS000716100MG

CyclopentY isothiocyana , 100mg
About this Item:
CAS #: 33522-03-1
MDL #: MFCD00040877
Molecular Weight: 127.21
UNSPSC Code: 12352200
Chemical Formula: C6H9NS
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.