Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentyl iodide, 98%, stabilized over copper, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
1556-18-9
C5H9I
196.03
PCEBAZIVZVIQEO-UHFFFAOYSA-N
cyclopentane, iodo,cyclopentyl iodide,jodcyclopentan,cyclopentyliodide,iodo-cyclopentane,iodocyclo-pentane,iodocyclopentane,acmc-1c0qu
73784
iodocyclopentane
C1CCC(C1)I
1556-18-9
C5H9I
196.03
PCEBAZIVZVIQEO-UHFFFAOYSA-N
cyclopentane, iodo,cyclopentyl iodide,jodcyclopentan,cyclopentyliodide,iodo-cyclopentane,iodocyclo-pentane,iodocyclopentane,acmc-1c0qu
73784
iodocyclopentane
C1CCC(C1)I
Color Colorless to Yellow
Flash Point 52°C
Specific Gravity 1.695
Infrared Spectrum Authentic
Boiling Point 166.0°C to 167.0°C
Stabilizer (Copper)
Density 1.6950g/mL
Quantity 5mL
Packaging Glass bottle
Assay Percent Range 96% min. (GC)
Linear Formula C5H9I
Formula Weight 196.03
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Cyclopentyl iodide, Stabilized
Show More Show Less

GHS H Statement
Flammable liquid and vapour.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If eye irritation persists: Get medical advice/a

GHS Signal Word: Warning

EINECSNumber : 216-311-1