Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentyl isocyanide, 96%, Acros Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
68498-54-4
C6H9N
95.14
XTLSDEJNOQVCHL-UHFFFAOYSA-N
cyclopentyl isocyanide,cyclopentylisocyanide,isocyano-cyclopentan,isocyano-cyclopentane,cyclopentyl isonitrile,3-isocyano-cyclopentane,acmc-1bgsw,tos-bb-0762
2734784
isocyanocyclopentane
[C-]#[N+]C1CCCC1
68498-54-4
C6H9N
95.14
XTLSDEJNOQVCHL-UHFFFAOYSA-N
cyclopentyl isocyanide,cyclopentylisocyanide,isocyano-cyclopentan,isocyano-cyclopentane,cyclopentyl isonitrile,3-isocyano-cyclopentane,acmc-1bgsw,tos-bb-0762
2734784
isocyanocyclopentane
[C-]#[N+]C1CCCC1
Boiling Point 40°C (11.0 hPa)
Infrared Spectrum Authentic
Density 0.8600g/mL
Specific Gravity 0.86
Color Colorless to Yellow
Quantity 5g
Packaging Glass bottle
Refractive Index 1.4390 to 1.4410
Assay Percent Range 95% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 95.14
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material Cyclopentyl isocyanide
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not eat,drink or smoke when using this product.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and

GHS Signal Word: Warning