Alfa Aesar™ Cyclopentyl thiocyanate, 95%

Manufacturer: Alfa Aesar™ H32230ME

Quantity 500mg

 View more versions of this product

Catalog No. AAH32230ME

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C6H9NS
    Formula Weight 127.21
    MDL Number MFCD11100558
    Chemical Name or Material Cyclopentyl thiocyanate

    Specifications

    Formula Weight 127.21
    Health Hazard 1 Warning
    Molecular Formula C6H9NS
    MDL Number MFCD11100558
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P280f
    Wear protective clothing.
    Boiling Point 84°C (8mmHg)
    Quantity 500mg
    Chemical Name or Material Cyclopentyl thiocyanate
    Assay 95%

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P280f
    Wear protective clothing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit