Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cyclopentylacetic acid, 97+%

Click to view available options
Quantity:
5g
25g
100g
1123-00-8
C7H12O2
128.171
MFCD00001387
YVHAIVPPUIZFBA-UHFFFAOYSA-N
cyclopentylacetic acid,cyclopentaneacetic acid,cycylopentylacetic acid,carboxymethyl cyclopentane,cyclopentyl acetic acid,cyclopentyacetic acid,cyclopentanacetic acid,cyclopentyl-acetic acid,2-cyclopentylaceticacid,cyclopentane acetic acid
71606
2-cyclopentylacetic acid
C1CCC(C1)CC(=O)O
1123-00-8
C7H12O2
128.171
MFCD00001387
YVHAIVPPUIZFBA-UHFFFAOYSA-N
cyclopentylacetic acid,cyclopentaneacetic acid,cycylopentylacetic acid,carboxymethyl cyclopentane,cyclopentyl acetic acid,cyclopentyacetic acid,cyclopentanacetic acid,cyclopentyl-acetic acid,2-cyclopentylaceticacid,cyclopentane acetic acid
71606
2-cyclopentylacetic acid
C1CCC(C1)CC(=O)O
Refractive Index 1.453
Density 1.023
Quantity 25g
Flash Point 114°C (237°F)
Boiling Point 231°C
Odor Rancid
Melting Point 12°C to 14°C
Beilstein 2040821
Formula Weight 128.17
Percent Purity ≥97%
Chemical Name or Material Cyclopentylacetic acid
Show More Show Less

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 214-368-7

TSCA : No

Recommended Storage : Ambient temperatures