Alfa Aesar™ Cyclopentylacetylene, 97%

Manufacturer: Alfa Aesar™ H5348306

Quantity 5g

 View more versions of this product

Catalog No. AAH5348306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 930-51-8
    Molecular Formula C7H10
    Formula Weight 94.16
    MDL Number MFCD00013744
    UN Number UN3295
    Synonym Ethynylcyclopentane
    Chemical Name or Material Cyclopentylacetylene

    Specifications

    Formula Weight 94.16
    Molecular Formula C7H10
    MDL Number MFCD00013744
    UN Number UN3295
    Synonym Ethynylcyclopentane
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H301
    Highly flammable liquid and vapor.
    Toxic if swallowed.
    Health Hazard 3 GHS P Statement
    P210-P280-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 1°C (34°F)
    Boiling Point 107° to 109°C
    Quantity 5g
    CAS 930-51-8
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
    Chemical Name or Material Cyclopentylacetylene
    Assay 97%

    Description

    Applications
    Cyclopentylacetylene is used as pharmaceutical intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H225-H301
    Highly flammable liquid and vapor.
    Toxic if swallowed.

    GHS P Statement
    P210-P280-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit