Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cyclopentylacetylene, 97%

Manufacturer:  Alfa Aesar™ H5348314

Quantity 25g

 View more versions of this product

Catalog No. AAH5348314


  Check Availability
Add to cart

Specifications

Description

Applications
Cyclopentylacetylene is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

930-51-8
94.16
UN3295
Cyclopentylacetylene
C7H10
MFCD00013744
Ethynylcyclopentane

Specifications

1°C (34°F)
25g
C7H10
MFCD00013744
Ethynylcyclopentane
97%
107° to 109°C
930-51-8
94.16
UN3295
Cyclopentylacetylene

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H301
Highly flammable liquid and vapor.
Toxic if swallowed.

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit