Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentylboronic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
63076-51-7
C5H11BO2
113.95
VTTDFSNKIMAQTB-UHFFFAOYSA-N
cyclopentaneboronic acid, boronic acid, cyclopentyl, cylopentyl boronic acid, cyclopentylboronicacid, pubchem7792, acmc-209ncu, --paclitaxel;paclitaxel, boronic acid, b-cyclopentyl, cyclopentylboronic acid
2734327
cyclopentylboronic acid
B(C1CCCC1)(O)O
63076-51-7
C5H11BO2
113.95
VTTDFSNKIMAQTB-UHFFFAOYSA-N
cyclopentaneboronic acid, boronic acid, cyclopentyl, cylopentyl boronic acid, cyclopentylboronicacid, pubchem7792, acmc-209ncu, --paclitaxel;paclitaxel, boronic acid, b-cyclopentyl, cyclopentylboronic acid
2734327
cyclopentylboronic acid
B(C1CCCC1)(O)O
Melting Point 134.0°C to 142.0°C
Infrared Spectrum Authentic
Color White
Quantity 10g
Assay Percent Range 95%
Formula Weight 113.95
Physical Form Crystalline Powder or Flakes
Percent Purity 95%
Chemical Name or Material Cyclopentylboronic acid

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning