Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclopentylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™, ACROS Organics™
Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
Specifications
Packaging | AcroSeal™ Glass Bottle |
Specific Gravity | 0.86 |
Quantity | 100mL |
Density | 0.8600g/mL |
Flash Point | −40°C |
Linear Formula | C5H9MgCl |
Formula Weight | 128.88 |
Chemical Name or Material | Cyclopentylmagnesium chloride, 2M solution in diethyl ether |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.
Extremely flammable liquid and vapor.
May form explosive
GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger