Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclophilin 40 Mouse anti-Human, Clone: 1B8, Novus Biologicals™

Mouse Monoclonal Antibody

$200.00 - $300.50

Specifications

Antigen Cyclophilin 40
Clone 1B8
Concentration 1 mg/mL
Dilution Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB60239100U View Documents Novus BiologicalsSupplier Diversity Partner
NBP260239100UL
100 ul Each for $300.50
Add to cart
 
NB6023950UL View Documents Novus BiologicalsSupplier Diversity Partner
NBP26023950UL
50 ul Each for $200.00
Add to cart
 
Description

Description

Cyclophilin 40 Monoclonal antibody specifically detects Cyclophilin 40 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Cyclophilin 40
1 mg/mL
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Purified
RUO
Human
40 kDa peptidyl-prolyl cis-trans isomerase, 40 kDa peptidyl-prolyl cis-trans isomerase D, cyclophilin D, Cyclophilin-40, Cyclophilin-related protein, CYP40, CYP-40cyclophilin 40, CYPD, EC 5.2.1.8, MGC33096, peptidyl-prolyl cis-trans isomerase D, peptidylprolyl isomerase D, peptidylprolyl isomerase D (cyclophilin D), PPIase D, Rotamase D
PPID
IgG2b Kappa
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1B8
Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100
Unconjugated
Mouse
Immunology
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
5481
Recombinant human PPID (1-370aa) purified from E. coli
Monoclonal
Protein A purified

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit